Nalgene® Staining Boxes, Thermo Scientific
Supplier: Thermo Fisher Scientific
5705-1010
28196-310CS 108.7 USD
28196-310
Nalgene® Staining Boxes, Thermo Scientific
Staining Boxes
Boxes are made of polymethylpentene and feature a polypropylene copolymer cover and a low-density polyethylene plug
Use box to stain, fix, destain, and handle electrophoresis gels and membranes with less chance of damage than conventional methods. Box 28196-310 holds up to 500mL of liquid when a gel is in place; box 28196-306 holds up to 750mL. Both box and cover will resist temperatures from –70 to 70°C (–94 to 158°F), and have good chemical resistance to acids, bases, and select organic solvents. Strategic plug location allows easy liquid drainage with minimal gel handling. The plug will not interfere with the cover or box contents.