Whatman™ Nylon Membrane Circles
Supplier: Cytiva
7402-004 7404-001 7404-002 7404-004 7404-009 7408-004 7402-009 7402-001 7402-002 7410-004
28159-723PK 204.25 USD
28159-723 28159-743 28159-744 28159-746 28159-751 28159-768 28159-728 28159-720 28159-721 10035-550
Whatman™ Nylon Membrane Circles
Filters Membrane Filters
High-quality nylon membranes are suitable for filtering aqueous solutions and most organic solvents. The membranes are suitable for use with a wide range of biological preparations and can be used where other membranes are unsuitable or difficult to use.
  • Flexible, durable and tear resistant
  • Can be autoclaved at 135 °C
Nylon membranes are hydrophilic, eliminating the need for wetting agents that could be extracted when filtering aqueous solutions.