Ultipor® Nylon Membrane Filter Discs, Cytiva (Formerly Pall Lab)
Supplier: Cytiva
NX047100
28148-028PK 178.51 USD
28148-028
Ultipor® Nylon Membrane Filter Discs, Cytiva (Formerly Pall Lab)
Filters Membrane Filters
Ultipor® Nylon Filter Discs are made of a reinforced single layer of N66 Nylon with nonwoven internal polyester.
  • Reinforced single layer
  • Used in fine filtrations
  • For small volumes, membrane area 5 cm2
These filter discs are used in sterilization, HPLC, and other fine filtrations of small volume solutions.