HEPA Capsule, Cytiva (Formerly Pall Lab)
Supplier: Cytiva
12144
28145-145EA 130.99 USD
28145-145
HEPA Capsule, Cytiva (Formerly Pall Lab)
Filters Capsule Filters
Designed to provide bacteria-free air when required for sterile applications.
  • Polypropylene housing
  • Acrylic copolymer membrane
  • High flow rates
  • Low differential pressures
Polypropylene housing contains Versapor® supported acrylic copolymer membrane with 1.2µm pore size. Provides high flow rates at low differential pressures. Not autoclavable.
Also for non sterile liquid prefiltration.