Mini Capsule Filter, 0.45 µm, Sterile, Cytiva (Formerly Pall Lab)
Supplier: Cytiva
12123
28145-112EA 143.29 USD
28145-112
Mini Capsule Filter, 0.45 µm, Sterile, Cytiva (Formerly Pall Lab)
Filters Capsule Filters
Use both for prefiltration of particulate liquids prior to final sterilizing filtration, and for removal of particles and debris from liquids not requiring final sterilization.
  • Two layers
  • Barbed hose fittings
  • Polycarbonate housing
Incorporates two layers of Versapor® (acrylic copolymer) membrane: 0.8µm prefilter layer followed by 0.45 µm final filter.
Polycarbonate housing is fitted with 12.7 mm (¹/₂") male, barbed, hose fittings for rapid installation with flexible tubing. Biosafe and disposable. Gamma irradiation sterilized.
Not autoclavable and not suited for dry-heat applications.