AcroVent® Filter Devices, Cytiva (Formerly Pall Lab)
Supplier: Cytiva
4249
28144-910PK 134.48 USD
28144-910
AcroVent® Filter Devices, Cytiva (Formerly Pall Lab)
Filters In-Line Filters
Disposable 50 mm filter devices for use in routine, noncritical venting applications.
  • Disposable
  • For routine venting applications
Ideal for gas lines for CO₂ incubators as they significantly decrease risk of contamination. The filter media is constructed of 0.2 µm PTFE on a polypropylene support in a polypropylene housing. The effective filtration area is 19.6cm2. Inlet and outlet connections are 6.4 to 12.7 mm (¹/₄ to ¹/₂") diameter stepped hose barbs.