In-Line Filter Holder, Stainless Steel, 47 mm, Cytiva (Formerly Pall Lab)
Supplier: Cytiva
2220
28144-506EA 1192.49 USD
28144-506
In-Line Filter Holder, Stainless Steel, 47 mm, Cytiva (Formerly Pall Lab)
Filter Holders
High-quality, corrosion-resistant in-line filter holder is designed for stack sampling.
  • Membrane or glass fiber filters
  • Holder opens and closes easily
  • Filter remains undisturbed for particulate analysis
  • Upstream threaded vent allows release of trapped gas during liquid filtration
Body, center ring collar, support screen, and underdrain disc are made of Type 316 stainless steel. Captive thrust ring is PTFE; O-ring is Viton® rubber; hose barb adapters are polyethylene. Inlet/outlet connections: 9.5 mm (3/8") FNPT. Hose barbs accept 6.4 mm (1/4") I.D. tubing. Autoclavable.