Acrodisc® Syringe Filters, DMSO-Safe, Sterile, Cytiva (Formerly Pall Lab)
Supplier: Cytiva
4433
28144-050PK 215.46 USD
28144-050
Acrodisc® Syringe Filters, DMSO-Safe, Sterile, Cytiva (Formerly Pall Lab)
Filters Syringe Filters
These 25mm filters are ideal for the sterilization of media used for cell cryopreservation.
  • Nylon
  • Polypropylene housing
  • Luer-lock inlet
Constructed of nylon filter media in a polypropylene housing, they are compatible with solutions containing DMSO. Filters feature a female Luer-lock inlet connection and a male slip Luer outlet connection, and operate at a maximum temperature of 55 °C (131 °F). Sterilized by ethylene oxide.
Ordering information: Individually blister packed.