Acrodisc® Syringe Filters, 4 mm, Cytiva (Formerly Pall Lab)
Supplier: Cytiva
4472
28143-978CS 1786.95 USD
28143-978 77603-476
Acrodisc® Syringe Filters, 4 mm, Cytiva (Formerly Pall Lab)
Filters Syringe Filters
These syringe filters are ideal for filtration of gas and organic solvents.
  • Polypropylene housing
  • HPLC certified
The polypropylene housing is fitted with female Luer-lock inlet and male slip Luer outlet. The hydrophobic PTFE membrane, on a polypropylene support, has broad chemical compatibility. The hydrophilic nylon membrane has excellent compatibility with esters, bases, and alcohols, and requires no prewetting.
They can also be used for air filtration and venting.
Certifications: Both membranes are HPLC certified for low levels of UV-absorbing extractables.