Acrodisc® PSF Syringe Filters, 25 mm, Cytiva (Formerly Pall Lab)
Supplier: Cytiva
AP-4549 AP-4787 AP-4788 AP-4792 AP-4308 AP-4522 AP-4225 AP-4521 AP-4302 AP-4517 AP-4518 AP-4790 AP-4791 AP-4793 AP-4796 AP-4797 AP-4309 AP-4436 AP-4520 AP-4219 AP-4438 AP-4794 AP-4500 AP-4528 AP-4408 AP-4310 AP-4303 AP-4501 AP-4502 AP-4548 AP-4910 AP-4912 AP-4913 AP-4914 AP-4915 AP-4916 AP-4917 AP-4918 AP-4919 AP-4920 AP-4921 AP-4438TC
28143-910CS 905.35 USD
28143-910 97027-938 97027-940 97027-948 16003-726 28143-278 28143-384 28143-392 28143-870 28143-901 28143-903 97027-944 97027-946 97027-950 97027-962 97027-964 16003-722 16003-728 28143-274 28143-390 28143-866 28143-869 28143-882 97027-956 16003-720 28143-912 16003-724 16003-730 28143-872 28143-884 28143-886 28143-915 76308-662 76308-664 76308-666 76308-668 76308-670 76308-672 76308-674 76308-676 76308-678 76308-680 76308-682 76308-684 77610-708
Acrodisc® PSF Syringe Filters, 25 mm, Cytiva (Formerly Pall Lab)
Filters Syringe Filters
The new Acrodisc® One and Acrodisc® PSF premium syringe filters combine the benefits of a multi-layered prefiltration device with an automation-certified filter housing.
  • Increased surface area for maximizing throughput
  • Heat sealed membrane for reduced breakthrough
  • Accurate analysis. HPLC certified for low levels of UV-absorbing extractables
  • Available in 0.2 µm for UHPLC applications and with GxF (Glass Fiber) pre-filter for difficult to filter samples
  • Automation Certified to ensure smooth operation and worry-free performance 24 hours a day in automated workstations
  • The Acrodisc® One offers a Universal Membrane for all your analytical filtration requirements
Polypropylene housing with female Luer inlet and male slip outlet connections.
Acrodisc ONE with water wettable PTFE (wwPTFE) has a Next Generation Universal Membrane for all sample types with fast flow rates, low protein binding, and low levels of UV-absorbing extractables. Avoid the expense and inconvenience of keeping a variety of filters on hand, it is a versatile filter for aqueous and aggressive organic solvent-based solutions.
Other Acrodisc® PSF filter membrane options:
PVDF membrane has excellent compatibility with aggressive acids and alcohols.
PES membrane is available in a special Ion Chromatography design or with a GxF prefilter.
Nylon membrane has excellent compatibility with esters, bases, and alcohols, and requires no prewetting.
Hydrophobic PTFE membrane offers exceptional chemical and temperature compatibility and is ideal for gas, organic solutions, air filtration, or venting.
The Acrodisc® One and Acrodisc® PSF GxF syringe filters provide two to four times the throughput of standard prefilter devices for particulate-laden samples.
For ion chromatography specific syringe filters, look for the Acrodisc® ion chromatography syringe filters page.
Packaging: Syringe filters packed in cases of 200 are available in four packs of 50 or in an AutoPack™ format. The AutoPack™ format consists of eight tubes with 25 syringe filters stacked on top of each other.