Whatman™ GD/X Syringe Filters, Prefilter, Non-Sterile, Whatman products (Cytiva)
Supplier: Cytiva
6905-2504 6891-2507 6870-1304 6872-1304 6874-1304 6882-1316 6870-2502 6870-2504 6872-2502 6872-2504 6874-2502 6874-2504 6876-2502 6876-2504 6880-2502 6880-2504 6884-2510 6890-2507 6894-2504 6871-2502 6871-2504 6875-2502 6875-2504 6870-2550 6881-2504 6895-2504 6894-1304 6874-1302 6871-1304 6888-2527 6870-1302 6872-1302 6873-1304 6882-2516 6892-2515 6873-2502 6873-2504 6876-1302 6886-2512 6905-2502 6887-2502 6882-2504 6883-2504 6871-2550 6883-2516 6884-1310 6875-1302 6880-1302 6888-1327 6890-1307
14217-554PK 4309.2 USD
14217-554 14223-314 28138-104 28138-108 28138-112 28138-124 28138-126 28138-154 28138-156 28138-158 28138-160 28138-162 28138-164 28138-166 28138-168 28138-172 28138-174 28138-176 28138-180 28138-186 28138-190 28138-192 28138-194 28138-200 28138-202 70240-004 80087-208 28138-170 28138-204 28138-138 28138-110 28138-120 28138-142 28138-150 28138-184 28138-102 28138-106 28138-118 28138-146 28138-178 28138-188 28138-196 28138-198 28138-114 28138-182 28138-140 89233-778 89233-780 89233-782 89233-784 10035-516 10035-518 10035-520 10035-522 10147-962 10147-966 10147-960 10147-964 10147-968 10147-970 10147-972 75831-558
Whatman™ GD/X Syringe Filters, Prefilter, Non-Sterile, Whatman products (Cytiva)
Filters Syringe Filters
Whatman GD/X Syringe Filters are high-viscosity filters that incorporate a prefilter to enable filtration of viscous, dense or otherwise hard-to-filter material with high solids content as in environmental and food analysis samples.
  • Recyclable after use
Non sterile Whatman GD/X Syringe Filters combine a polypropylene housing, high-efficiency Whatman GMF 150 and GF/F pre-filter media, and one of the following membrane materials:
- Polyethersulfone (PES) for serum, plasma and tissue culture
- Polyvinylidene difluoride (PVDF) for HPLC
- Cellulose acetate for biological solutions
- Borosilicate glass microfiber
The prefilter contains a high-efficiency borosilicate glass filtration stack that will retain fine particles down to 0.7 μm, allowing difficult sample filtration with less hand force. It reduces the need for multiple syringe filters by processing three to seven times more sample volume than syringe filters without prefilters. The filters can be autoclaved.
Cytiva also offers Sterile Whatman GD/X syringe filters.