Whatman™ Polycap™ TF Disposable Filter Capsules, Whatman products (Cytiva)
Supplier: Cytiva
6710-3602 6700-3602 6700-3610 6700-7502 6700-7504 6701-7510 2702T 6710-7502 2801 2601T 2602S 2603T 2700T 2702M 2703T 2802T 2803T 2804T 6711-3604
28137-708EA 118.05 USD
28137-708 28137-722 28137-728 28137-872 28137-874 28137-876 28137-878 28137-880 28137-882 89013-354 28137-716 28137-720 97059-550 10035-352 10035-354 10035-356 10035-370 10035-372 10035-374 10035-406 10035-408 10035-410 10035-412 10147-952
Whatman™ Polycap™ TF Disposable Filter Capsules, Whatman products (Cytiva)
Filters Capsule Filters
Cytiva's Whatman Polycap TF filters are made with durable, hydrophobic polytetrafluoroethylene (PTFE) membranes in a polypropylene housing for use with organic solvents and chemically aggressive solutions.
  • PTFE filter membrane is resistant to most solvents
  • Available in 0.05, 0.1, 0.2, 0.45, and 1.0 µm pore sizes
  • 0.05 µm pore size capsules capsules designed for ultra-clean applications
  • 1.0 µm pore size capsules used for extended life and filtration of highly contaminated solutions
  • Autoclavable capsule filtration device if sterility is required
Filtration of organic solvents and aggressive solutions requires a durable membrane for filtration without sample contamination.
Autoclavable Polycap TF capsule filters feature PTFE membranes with high chemical resistance. Common applications include venting, in-line filtration to sterilize and remove particles from air/gas/liquid streams, filtration to ensure clean inert gases in vial filling lines, and isolation of exhaust from fermenters.
Polycap TF filters support use in the electronics, pharmaceutical, and biopharmaceutical industries, because they are photoresistant and can be used to vent and perform in-line filtration for production systems in biological environments.
Certifications: Manufactured according to ISO9001:2008 QMS standards