WHEATON® Filtration Assembly with Fritted Glass Support and No. 8 Stopper Connections, 47 mm, DWK Life Sciences
Supplier: DWK Life Sciences (KIMBLE)
419347
27589-098CS 374.1 USD
27589-098
WHEATON® Filtration Assembly with Fritted Glass Support and No. 8 Stopper Connections, 47 mm, DWK Life Sciences
Funnels Filtration Funnels Filtration Apparatus Glass
The filtration assembly with fritted glass support is designed to handle up to 1000 ml of liquid for analysis of particulate or microbiological contamination.
  • Glass components are manufactured from WHEATON 33 low extractable borosilicate glass
  • Recommended for general filtration
Flasks have a #2 hose connection for 1/4” (6 mm) I.D. tubing. Flasks are not included.
Certifications: Glass conforms to USP Type I and ASTM E 438 Type I, Class A requirements.
Delivery information: Includes a coarse porosity (40 - 60 μm) fritted glass support base, 300 ml graduated funnel, anodized aluminum clamp, and a No. 8 silicone stopper.