WHEATON® Filtration Assembly with Stainless Steel Support and No. 5 Stopper Connections, 25 mm, DWK Life Sciences
Supplier: DWK Life Sciences (KIMBLE)
419327
27589-080CS 436.85 USD
27589-080
WHEATON® Filtration Assembly with Stainless Steel Support and No. 5 Stopper Connections, 25 mm, DWK Life Sciences
Funnels Filtration Funnels Filtration Apparatus Glass
The filtration assembly with fritted glass support is designed to handle small volumes of liquids for analysis of particulate or microbiological contamination.
  • Glass components are manufactured from WHEATON 33 low extractable borosilicate glass
  • Recommended for filtering viscous or proteinaceous solutions
  • or to produce ultraclean filtrate
Flasks have a #2 hose connection for 1/4” (6 mm) I.D. tubing. Flasks are not included.
Certifications: Glass conforms to USP Type I and ASTM E 438 Type I, Class A requirements.
Delivery information: Includes a 316 stainless steel support with 120 mesh screen, PTFE support screen gasket, glass support base, 15 ml graduated funnel, anodized aluminum clamp and a No. 5 silicone stopper.