BioTrace™ NT Nitrocellulose Transfer Membranes, Cytiva (Formerly Pall Lab)
Supplier: Cytiva
66485
27377-000PK 295.3 USD
27377-000 28139-306 27376-991
BioTrace™ NT Nitrocellulose Transfer Membranes, Cytiva (Formerly Pall Lab)
Transfer Membranes
Pure unsupported nitrocellulose membranes have high binding affinity (209 µg/cm2) and are colony/plaque lifts and protein transfers.
  • Strong and durable, less likely to tear or crack than competitor nitrocellulose
  • High binding capacity for proteins and nucleic acids
  • Lower protein burn-through than competitors in electrophoretic transfers
  • Nonsterile
BioTrace™ NT provides a good binding matrix for macromolecules. The unique structure and lack of growth-inhibiting ingredients facilitate bacterial growth even when the bacteria are stressed by transformation. Typical thickness: 140 µm.
Applications include protein transfers, western blots, southern blots, northern blots, and nucleic acid and protein dot blots.