Owl™ Double Sided Gel Vertical Electrophoresis System, Model P82, Thermo Scientific
Supplier: Thermo Fisher Scientific
P82
27373-566EA 1228.2 USD
27373-566
Owl™ Double Sided Gel Vertical Electrophoresis System, Model P82, Thermo Scientific
Electrophoresis Systems Vertical Gel Electrophoresis Systems
For flat, even banding and crisp resolution
System includes an upper buffer chamber, lower buffer chamber, SuperSafe™ lid with attached power supply leads, two 1.3cm (1/2") thick wedges, and two 1.6 cm (5/8") thick wedges. The two sizes of wedges are provided to accommodate varying thickness of precast gels.
The system can run two 10 W x 10 L cm gels, or a single gel using the blocking plate. The system can accommodate 10 W x 8 L cm (4 x 31/8") gels utilizing the provided adapter. Gels can be poured or precast. The system is designed to allow gels to be easily placed into the chamber. A wedge is placed in front of the cassette to provide even pressure against the leakproof gasket and ensure that the cassette is held in the proper running position. A molded gasket provides a leak-resistant seal. The lid closes in one orientation only, ensuring that the positive and negative electrodes are not reversed. No cooling is required.
Accessories information: Combs are available in standard or preparative types.