Owl™ Gel Staining Box, Thermo Scientific
Supplier: Thermo Fisher Scientific
GSB-3
27373-406PK 78.92 USD
27373-406
Owl™ Gel Staining Box, Thermo Scientific
Staining Boxes
Thermo Scientific™ Owl Gel Staining Boxes provide a convenient, conventional staining method
Stain, fix and de-stain in the same container. Each container will hold up to 125 mL of liquid and one gel when used on a shaking platform. Grooved lid enables proper orientation and stability when stacking. Made of clear plastic, so gels can be viewed directly in the tray.