Owl™ Micro Electrophoresis System, Model C2, Thermo Scientific
Supplier: Thermo Fisher Scientific
C2-S
27373-322EA 390.71 USD
27373-322
Owl™ Micro Electrophoresis System, Model C2, Thermo Scientific
Electrophoresis Systems Horizontal Gel Electrophoresis Systems
Horizontal microgel system is designed for quick restriction fragment analysis
Glass slides measuring 2x3" (5x7.5cm) serve as the gel tray. To use, pour and form thin gels by surface tension directly on glass slide; complete run lasts five to 10 minutes. Using available caster and casting slides, gels can be cast and solidified in less than 10 minutes.
Microgel may also be used to quickly check a few PCR samples before loading a large number of samples on a larger gel, saving time.
Ordering information: System includes buffer chamber (100mL buffer volume), lid, power supply leads, casting tray, 10 casting slides, and one 1.5mm thick, 7-well comb.