Owl™ EasyCast™ Gel System with Built-in Recirculation, Models A5 and B3, Thermo Scientific
Supplier: Thermo Fisher Scientific
A5
27372-134EA 1326.1 USD
27372-134 27372-292
Owl™ EasyCast™ Gel System with Built-in Recirculation, Models A5 and B3, Thermo Scientific
Electrophoresis Systems Horizontal Gel Electrophoresis Systems
Ideal for long runs, multiple sample sets, or RNA gels.
  • Circulation system is built into the gel chamber
  • No external pumps, tubing, stir bars or additional assembly is required
Bubbles are collected at the cathode end of the unit and shunted through a conduit tube to the anode end. The bubbles displace the buffer, creating effective recirculation within the chamber.
The self-recirculation system prevents the formation of pH and ionic gradients for high resolution and uniform, reproducible results. Eliminates uneven migration, band distortions, or dissociation of pH-dependent glyoxylated RNA molecules that can result when ionic depletion occurs.
Ordering information: The complete mini gel system includes a buffer chamber; SuperSafe™ lid with attached power supply leads; an EasyCast™ gasketed U.V. transmissible (UVT) gel tray; and two double-sided combs, one 12-well 1.5 mm thick and one 20-well 1 mm thick. The complete large gel system includes a buffer chamber; SuperSafe™ lid with attached power supply leads; UVT gel tray with gasketed endgates; three-point leveling base; and three 1.5 mm thick combs (16-, 24-, and 36-well).