Owl™ EC-105 Compact Power Supply, Thermo Scientific
Supplier: Thermo Scientific
105ECA-LVD
27370-266EA 1161.94 USD
27370-266
Owl™ EC-105 Compact Power Supply, Thermo Scientific
Power Supplies
Carry out DNA and RNA electrophoresis with the help of the Owl EC-105 Compact Power Supply, equipped with an overcurrent detector for maximum safety.
  • Easy-to-use power supply operates in constant voltage
  • Adjustment knob and hi-lo range switch control the output, which is variable in 10 V increments from 20 to 250 V
  • Two-position switch toggles the bright LED display from indicating output voltage to output current
  • Supply is equipped with an overcurrent detector for maximum safety and has two jacks