Oxygen/Natural Gas Sealing Torch, Labconco®
Supplier: Labconco
7578500
26675-851EA 1440 USD
26675-851
Oxygen/Natural Gas Sealing Torch, Labconco®
Heaters Gas Burners
Oxygen/Natural Gas Sealing Torch, specifically designed for flame sealing freeze dry ampules
Seals all types of heat-resistant gas. Connects to natural gas, butane or propane and oxygen with 1/4" ID hose connectors.