PYREX® Capillary Tip Bleed Tube, Corning
Supplier: Corning
7815-19
26658-037CS 18.63 USD
26658-037
PYREX® Capillary Tip Bleed Tube, Corning
Tubes Gas Inlet Tubes
For bleeding gases, steam, or liquids below the liquid surface of sidearm distillation flasks.