Chemware® PTFE Boiling Stones, Saint-Gobain Performance Plastics
Supplier: Saint Gobain Performance Plastics
D1069103
26397-103EA 272.75 USD
26397-103
Chemware® PTFE Boiling Stones, Saint-Gobain Performance Plastics
Boiling Stones
Ultra-pure, virgin PTFE chips.
  • promote gentle, efficient boiling by creating many projections on which small bubbles can form
  • Unaffected by boiling acids, hydrocarbons, ketones, esters, alcohols, or alkalies in any concentrations
  • Suitable for column packing
Ideal for continuous extraction methods, such as EPA SW846.
Packaging: Supplied in a 450 g bottle.