PYREX® Mallory Direct Reading Settlometer, Corning
Supplier: Corning
7250-2L
26307-604CS 440 USD
26307-604
PYREX® Mallory Direct Reading Settlometer, Corning
Funnels Sedimentation Cones
Used with oxidized sludge process for treatment of sewage and other wastes. These cylinders are graduated in cc per litre and in hundredths of a foot.
Int.Ø×H: 130×190.5 mm