Bard-Parker® Scalpel Handle, No. 6, Aspen Surgical
Supplier: Aspen Surgical Products
371060
25856-045CS 187.57 USD
25856-045
Bard-Parker® Scalpel Handle, No. 6, Aspen Surgical
Scalpels and Scalpel Assemblies Scalpel Handles
For use with No
  • Reusable
  • Nonsterile
  • No natural rubber latex
20, 21, 22, 23, 24, or 25 Bard-Parker® conventional blades. Ideally suited for tissue separation and other procedures that require a sharp surgical blade to puncture or cut.