Bard-Parker® Scalpel Handle, No. 5, Aspen Surgical
Supplier: Aspen Surgical Products
371050
25856-023CS 205.49 USD
25856-023
Bard-Parker® Scalpel Handle, No. 5, Aspen Surgical
Scalpels and Scalpel Assemblies Scalpel Handles
This durable handle accommodates No
10, 11, 12, or 15 Bard-Parker® Rib-Back™ blades. Handle is shaped to provide a firm grip for entire palm of hand.