Bard-Parker® Scalpel Handle, No. 7, Aspen Surgical
Supplier: Aspen Surgical Products
371070
25856-001CS 201.69 USD
25856-001
Bard-Parker® Scalpel Handle, No. 7, Aspen Surgical
Scalpels and Scalpel Assemblies Scalpel Handles
For use with No
10, 11, 12, and 15 Bard-Parker® Rib-Back™ blades. Narrower and of greater length than the No. 3 handle. Particularly efficient for delicate technique. Convenient for work in difficult positions to reach.