Dissecting Forceps, Fine Tip, Serrated, Grobet USA
Supplier: Grobet
57.929
25718-012EA 27.69 USD
25718-012
Dissecting Forceps, Fine Tip, Serrated, Grobet USA
Forceps
Made of nickel-plated, polished steel with fine, straight points having fine file-cut serrations on inner side
Provided with guide pin.