VWR® Staining Jar with Cover, Hellendhal Type
Supplier: VWR International
25460-907
25460-907CS 141.63 USD
25460-907
VWR® Staining Jar with Cover, Hellendhal Type
Jars Staining Jars
This tapered jar is made of optically clear PMP (polymethylpentene).
  • Optically clear
  • Polymethylpentene construction
  • Designed to accommodate multiple slides back to back
  • Cover included
It holds standard 76x26 mm (3x1") slides vertically. Eight grooves accommodate 16 slides back to back. Supplied with PMP cover. Xylene can cause etching after two days at room temperature.