Staining Dish, 10 Slide Unit, Wheaton
Supplier: DWK Life Sciences (KIMBLE)
900170
25455-000CS 350.37 USD
25455-000
Staining Dish, 10 Slide Unit, Wheaton
Jars Staining Jars
Designed for staining 3x1" (75x25mm) microscope slides
Holds 10 single slides, 19 slides arranged alternately straight across and diagonally, or 20 slides back-to-back. The dish and cover are manufactured from soda-lime glass. Approximate inside dimensions are 80Lx55Wx35D mm.