STAR™ DISH Space & Media Saver' Petri Dishes, Phoenix Biomedical'
Supplier: Phoenix Biomedical
390
25389-038CS 321.31 USD
25389-038
STAR™ DISH Space & Media Saver' Petri Dishes, Phoenix Biomedical'
Culture Dishes Petri Dishes
These space-saving petri dishes are specifically designed for commercial media pourers
The height of these dishes also saves storage space in the refrigerator or incubator for the end user. In addition to refrigerator and incubator storage space, the smaller diameter option is perfect for users who want to save even more on the amount of media poured.
The innovative design of these dishes decreases the amount of media necessary per plate. They also extend the shelf life and eliminate drying and shrinking of media.