STAR™ DISH ALPINE Petri Dishes, Two Sections, Phoenix
Supplier: Phoenix Biomedical
372
25389-034CS 315.19 USD
25389-034
STAR™ DISH ALPINE Petri Dishes, Two Sections, Phoenix
Culture Dishes Petri Dishes
These petri dishes are ideal when a larger surface is required for culture or antibiotic sensitivity discs
These dishes are ideal for simultaneous differential studies of organisms or media in a common environment. Costs are reduced by minimizing the use of dishes, media and space.