STAR™ DISH Sectional Petri Dishes, Phoenix Biomedical
Supplier: Phoenix Biomedical
332
25389-018CS 336.85 USD
25389-018
STAR™ DISH Sectional Petri Dishes, Phoenix Biomedical
Culture Dishes Petri Dishes
These sectional petri dishes are ideal for simultaneous differential studies of organisms or media in a common environment
Individual compartments are numbered for easy identification. Costs are reduced by minimizing the use of dishes, media and space.