SP Bel-Art Desi Can™ Reusable Desiccant Canister, Bel-Art Products, a part of SP
Supplier: Bel-Art Products, a Part of SP
F42046-0000
24988-350EA 36.26 USD
24988-350
SP Bel-Art Desi Can™ Reusable Desiccant Canister, Bel-Art Products, a part of SP
Desiccators Desiccants
Silica gel effectively absorbs moisture through perforations in the aluminum case.
  • Reusable design
  • Monitor changes color with saturation level
  • Unlimited potential life
One canister of Reusable Desi-Can™ Canister maintains a safe level of humidity in 0.09 cubic meter (3 cu. ft.) of space.
Excellent for use in closets, containers, tool boxes, camera cases and other storage areas to prevent rust and moisture.
It can be regenerated in a 150 ºC (300 ºF) oven in about 3 hours for unlimited life. A monitor window shows blue when recharged and pink when saturated.