SP Bel-Art Dry-Keeper™ Stacking Desiccator Cabinet, Bel-Art Products, a part of SP
Supplier: Bel-Art Products, a Part of SP
H42053-0001 H42053-0001
24983-331EA 912.42 USD
24983-331 490012-264
SP Bel-Art Dry-Keeper™ Stacking Desiccator Cabinet, Bel-Art Products, a part of SP
Desiccators Desiccator Cabinets
Perforated shelves are movable to suit your dry storage needs in this desiccator cabinet that stacks up to 5 units high.
  • Polystyrene construction
  • Adjustable, perforated shelves
  • Door disappears beneath cabinet when open
This smoke-tinted polystyrene cabinet has two removable shelves that can be set at four heights.
The pull-down door slides out of the way into a compartment located underneath the cabinet.
Ordering information: Includes a small desiccant tray.