Desiccators, Static-Free, Stainless Steel, Boekel Scientific
Supplier: Boekel
134900
24981-012EA 4110.45 USD
24981-012
Desiccators, Static-Free, Stainless Steel, Boekel Scientific
Desiccators Desiccators, Glass
The Boekel Scientific Statnip™ Desiccator line is designed for storing your ESD sensitive devices such as CMOS, MOSFETs, Hybrid Circuits and any static samples, under an inert clean and dry atmosphere
The stainless steel design with glass sides and a glass front door panel provides easy viewing of your samples while the inside glass surface has a vapor deposited film of tin oxide. These units can be stacked to preserve space while the shelf spacing can be adjusted in 3/4" increments.
The explanded stainless steel shelves glide on conductive runners and are grounded to stainless steel shelf supports, frame and grounding lug and are constructed to prevent static buildup. Less leaky than conventional plastic desiccators there is no need to purchase ionizers or apply topical antistats.
Ordering information: Units supplied with two shelf assemblies, 1/4" OD inlet port, grounding lug, instruction manual and Boekel Scientific's two-year warranty on parts and labor.