PYREX® Economy Double Metric Scale Cylinders, To Contain, Corning
Supplier: Corning
3025-1L 3025-2L 3025-100 3025-10 3025-250 3025-25 3025-500 3025-50
24709-669CS 124.7 USD
24709-669 24709-680 89090-508 89090-510 89090-512 89090-514 89090-516 89090-518
PYREX® Economy Double Metric Scale Cylinders, To Contain, Corning
Cylinders Measuring Cylinders
Suitable for schools and labs requiring general accuracy only.
The double metric scale includes one reading down from top line and the other reading from bottom of cylinder to top line. Reduced tendency to tip due to relatively short height. Bumper guards not included.
Ordering information: Bumper guards available separately. Contact your sales representative.