KIMAX® Culture Tubes, Disposable, with Screw Thread Finish, Kimble Chase
Supplier: DWK Life Sciences (KIMBLE)
73750-16125 73750-13100 73750-16100 73750-16150 73750-20125 73750-20150
24631-446CS 716.32 USD
24631-446 89003-568 89003-570 89003-572 89003-574 89003-576
KIMAX® Culture Tubes, Disposable, with Screw Thread Finish, Kimble Chase
Tubes Culture Tubes
Tubes are suitable for tissue culture work and general bacteriological use.
Tubes are made from KIMAX-51® borosilicate glass in compliance with ASTM E438, Type I, Class A or B glass requirements. Each tube has a chevron marking area. Tubes are shrink-wrapped in plastic film in modular trays. Supplied without caps.