Stainless Steel Perforated Table, Bandy
Supplier: Bandy
TBL60024
24450-012EA 6949.63 USD
24450-012
Stainless Steel Perforated Table, Bandy
Furniture Tables and Workbenches
Perforated stainless steel table with 1.9 cm (³/₄") hole pattern allows approximately 40% air flow.
  • Constructed of 16-gauge
  • Type 304 stainless steel
  • Completely electropolished
Bull nose front edge for worker safety and comfort. Adjustable leveling feet.
Ordering information: For optional drawer and CPU stand (shown), contact your Avantor sales representative.