Nunc® CryoFlex™ Tubing, Thermo Scientific
Supplier: Thermo Fisher Scientific
343958
24270-013CS 3286.28 USD
24270-013
Nunc® CryoFlex™ Tubing, Thermo Scientific
Cryogenic Canes
Flexible polyethylene tubing provides extra safety when freezing hazardous samples or storing in liquid nitrogen
Can be used as a cane to lower CryoTube™ vials into liquid nitrogen.