Vibratory Pulverizer, Bico Braun International
Supplier: Bico
VP-1989
24253-010EA 44309.14 USD
24253-010
Vibratory Pulverizer, Bico Braun International
Mills
For efficient grinding of production samples
  • Starting switch and timer
  • Insulated cabinet for reduced operating noise
  • 250 mL bowl capacity
  • 1 set of grinding components
  • 1 bowl and cover
The Vibratory Pulverizer uses two rings and a puck inside an 8" hardened chrome allow steel grinding bowl. A specially designed motor directly coupled to a rotating eccentric swings the bowl and components at a precise speed and distance for maximum grinding efficiency. Rapidly reducing a 1/2" (13 mm) wet or dry sample of mineral, ore, rock, soil and similar materials to a fine mesh of 400+.
Ordering information: Available options include capacities of 100GM, 150GM, and 250ML, and chrome alloy and tungsten carbide materials. Contact your sales representative for more information.