Pulverizer, Type UA, Bico™
Supplier: Bico
242-53
24253-000EA 29756.39 USD
24253-000
Pulverizer, Type UA, Bico™
Mills
Pulverizer features a straight drive, V-belt without gears
Note: With quartz ore previously crushed to 1/4", unit will produce 150 to 200-mesh powder at rate of 1lb./min.
Reduces with one grinding material 1/4" and smaller to desired mesh. Adjusting screw adjusts grinding plates for desired mesh. Grinding chamber opens easily for cleaning. Special grooved pulley, 2 hp enclosed motor with starting switch. Motor is 2hp, 3 phase.
Ordering information: Supplied with one iron alloy stationary plate and one iron alloy revolving plate. Specify voltage when ordering. Specify serial number of pulverizer when ordering drop clutch.