Herman Inquarts,, pellets
Supplier: BICO INC.
239-23
23923-006PK
386.01
USD
23923-006
Herman Inquarts,, pellets
Inquarts, Herman
Herman lnquarts (Gold Free) are little pellets of lead bullion, each containing a definite amount of silver.
Herman Inquarts are uniform in silver content, easy to handle, save time and insure results. The exact silver content is marked on each package. Stock packages average 2mq of silver.