Wide Form Crucibles, CoorsTek
Supplier: Coors Ceramics
60133
23815-161CS 847.64 USD
23815-161 23815-081
Wide Form Crucibles, CoorsTek
Crucibles
Wide form crucibles are glazed inside and out, except for the exterior bottom and rim.
These porcelain vessels can be used at temperatures up to 1150 °C (2102 °F). Crucibles can be supplied with a permanent numeral marked on the outside surface for an additional charge. Quotations for numbering are available upon request.