VWR® Dual Channel Thermometer
Supplier: VWR International
23609-230
23609-230EA 363.21 USD
23609-230
VWR® Dual Channel Thermometer
Thermometers Digital Thermometers
Ideal for measuring temperatures in petroleum products or wastewater and for detecting temperature variations in incubators, ovens, and baths.
  • Traceable®
  • Dual probes
Rugged case is chemical- and shock-resistant ABS plastic with a handy flip-open bench stand for easy monitoring
Thermometer senses the changes between inlet and outlet temperatures in any system setup. Supplied with two immediate-response Type-K thermocouple probes that show temperature changes in less than a second. The 1.9 cm (3/4") display shows either probe temperature or the difference in temperature between probes. Readable from 4.5m (15') away. Hold switch allows display to capture readings.
All Type-K probes may be used with liquids, air/gas, and semi-solids. Stainless steel probe has a 22.9 cm (9") stem with a 3mm (1/8") diameter and a 102 cm (401/8") cable. Beaded probe is 1.5mm (1/16") in diameter and features a 1.2m (4') fluoropolymer resin cable. The low-temperature stainless steel probe has a diameter of 4.3mm (3/16") and features a 30.5 cm (12") stem with a 12.7 cm (5") handle. Low-temperature probe is supplied with a 2.4m (8') expandable cable.
Uses all Type-K thermocouple probes.
Certifications: A serial numbered certificate is provided from an ISO 17025 calibration laboratory accredited by A2LA to indicate thermometer traceability to standards provided by the National Institute of Standards and Technology.
Ordering information: Supplied with two fast-response Type-K beaded probes (61161-372) and a 9V battery.