VWR® Traceable® Key-Chain Thermometer
Supplier: VWR International
23609-178
23609-178EA 73.97 USD
23609-178
VWR® Traceable® Key-Chain Thermometer
Thermometers Digital Thermometers
Perfect for carrying with you — clip it on a lab coat buttonhole, jacket zipper, or belt loop.
  • Traceable®
  • One-second response time
  • Portable temperature monitoring
  • Ensure proper experiment temperature
Carry the keys you can’t do without, and be assured your experiment is conducted at the right temperature. The 2.5 cm (1") stainless steel probe is designed for fast liquid measurements and penetration of semi-solids. Accurate readings when immersed only 0.87 cm (1/3"). Accuracy to ±1°C. Light weight and miniature size make it the convenient pocket thermometer.
Certifications: A serial numbered certificate is provided from an ISO 17025 calibration laboratory accredited by A2LA to indicate instrument traceability to standards provided by the National Institute of Standards and Technology.
Ordering information: Supplied with replaceable battery power unit for one year of continuous operation.