VWR® Digital Surface Thermometer
Supplier: VWR International
23609-172
23609-172EA 120.73 USD
23609-172
VWR® Digital Surface Thermometer
Thermometers Digital Thermometers
This unit incorporates a 13.3 cm (51/4") long stainless steel probe with a 1.3 cm (1/2") diameter metal disk at the end of the probe for surface temperature measurements.
  • Traceable®
  • Operates for over a year on one silver-oxide battery
Constructed of plastic and stainless steel, this unit is safer to use than a mercury thermometer
Unit is switchable between °C and °F, providing results in both units without conversion factors. Readings are updated every second.
Extra-large, bright LCD (liquid crystal display) is 1 cm (3/8") high.
Certifications: A serial numbered certificate is provided from an ISO 17025 calibration laboratory accredited by A2LA to indicate instrument traceability to standards provided by the National Institute of Standards and Technology.