VWR® Optik-Wipes™ Tissues
Supplier: VWR International
90027-10
23609-168PK 39.82 USD
23609-168
VWR® Optik-Wipes™ Tissues
Wipes Disposable Wipes and Towels Lens Papers
Designed specifically for wiping eyeglasses, goggles, computer screens, gauge windows, plastic glasses, fume hood windows, optical parts, and microscopes/magnifiers.
  • Individually dispensed
  • Strong, cleanroom-grade cloth is soft and lint-free
  • Clean and polish with one swipe
  • Works equally well with glass and all plastics
Constructed of cellulose and polyester, wipes will not scratch surfaces or fall apart. Recloseable container eliminates contamination from dust and dirt.
Premoistened cloths easily remove oil, grease, and lint. Non-woven cloth is premoistened with a unique formulation.
Ordering information: Individual pop-up 15.2×22.9 cm (6×9") wipes are packaged 100 per polyethylene canister.