Brass Cork Borer Sets, Boekel Scientific
Supplier: Boekel
1601AC
23457-060EA 336.84 USD
23457-060
Brass Cork Borer Sets, Boekel Scientific
Cork Borers
Made of brass tubing with strong permanent handles
The cork borer sharpener consists of a solid brass cone with a curved, steel knife blade securely mounted in a wooden handle. It is constructed so that the cutting edge of the borer is trimmed to the correct angle regardless of the borer size. Total length: 19cm (71/2"). For borers with tubes up to 25.4mm (1") O.D.
Cork borer set consists of sequentially numbered, nested tubes that start at 4.5mm (3/16") O.D. and gradually increase in size. A steel punch is included with each set.