VWR® Traceable® Double Thermometer
Supplier: VWR International
23226-656
23226-656EA 394.95 USD
23226-656
VWR® Traceable® Double Thermometer
Thermometers Digital Thermometers
Thermometer has dual channels that permit monitoring via two separate probes.
  • Traceable®
  • Dual channels
  • Dual display
  • Reads °C and °F
  • Memory recalls maximum and minimum readings
Dual display shows the temperature of one probe or the temperature of both probes, and the difference between probes (T1–T2)
Thermometer accepts all Type-K thermocouple probes. Type-K probes may be used with liquids, air/gas, and semi-solids. Stainless steel probe has a 22.9 cm (9") stem with a 3mm (1/8") diameter attached to a 102 cm (401/8") cable. Beaded probe is 1.5mm (1/16") in diameter and features a 1.2m (4') fluoropolymer resin cable. The low-temperature stainless steel probe has a diameter of 4.3mm (3/16") and features a 30.5 cm (12") stem with a 12.7 cm (5") handle. Low-temperature probe is supplied with a 2.4m (8') expandable cable. High-temperature probe has a 3m (10') braided metal wire cable, a smooth tip, and a diameter of 5mm (3/16").
Display is 3.5 cm (13/8") high. Memory recalls maximum and minimum readings, and hold button freezes display so that readings can be captured and recorded. Serial RS-232 output allows the thermometer to be connected to a computer or data logger for capturing and keeping results.
Certifications: A serial numbered certificate is provided from an ISO 17025 calibration laboratory accredited by A2LA to indicate thermometer traceability to standards provided by the National Institute of Standards and Technology.
Ordering information: Supplied with two fast response Type-K beaded probes (61161-372) and a 9V battery.